missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OBFC1, Mouse anti-Human, Clone: 3G12-1B7, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00079991-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: MQPGSSRCEEETPSLLWGLDPVFLAFAKLYIRDILDMKESRQVPGVFLYNGHPIKQVDVLGTVIGVRERDAFYSYGVDDSTGVINCICWKKLNTESVSAAPSAARELSLTSQLKKLQETIEQKTKIEIGDTIRVRGSIRTYREEREIHATAYYKVDDPVWNIQIARMLELPTIYRKVYDQPFHSSALEKEEALSNPGALDLPSLTSLLSEKAKEFLMENRVQSFYQQELEMVESLLSLANQPVIHSACSDQVNFKKDTTSKAIHSIFKNAIQLLQEKGLVFQKDDGFDNLYYVTREDKDLHRKIHRIIQQDCQKPNHMEKGCHFLHILACARLSIRPGLSEAVLQQVLELLEDQSDIVSTMEHYYTAFSpecifications
| OBFC1 | |
| Monoclonal | |
| Unconjugated | |
| oligonucleotide/oligosaccharide-binding fold containing 1 | |
| AAF-44/FLJ22559/RP11-541N10.2/RPA-32/bA541N10.2 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 3G12-1B7 | |
| In 1x PBS, pH 7.4 | |
| BC017400 | |
| OBFC1 | |
| OBFC1 (AAH17400, 1 a.a. ∼ 368 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 79991 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction