missing translation for 'onlineSavingsMsg'
Learn More

Obefazimod, MedChemExpress

Product Code. 16485927
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485927 5 mg 5mg
16469746 50 mg 50mg
16464476 100 mg 100mg
16463198 10 mg 10mg
16462666 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16485927 Supplier Medchem Express Supplier No. HY100870
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Obefazimod (ABX464) is a potent anti-HIV agent. Obefazimod inhibits HIV-1 replication in stimulated peripheral blood mononuclear cells (PBMCs) with an IC50 ranging between 0.1 μM and 0.5 μM.

Obefazimod (ABX464) is a potent anti-HIV agent. Obefazimod inhibits HIV-1 replication in stimulated peripheral blood mononuclear cells (PBMCs) with an IC50 ranging between 0.1 μM and 0.5 μM.

Specifications

Chemical Name or Material Obefazimod
CAS 1258453-75-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H10ClF3N2O
Quantity 5 mg
Synonym ABX464
Solubility Information DMSO : ≥ 100 mg/mL (295.24 mM)
SMILES FC(F)(F)OC1=CC=C(NC2=NC3=C(Cl)C=CC=C3C=C2)C=C1
Molecular Weight (g/mol) 338.71
Formula Weight 338.71
Percent Purity 99.98%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.