missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
OBCAM/OPCML Polyclonal specifically detects OBCAM/OPCML in Rat samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | OBCAM/OPCML |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | IgLON family member 1, IGLON1opioid-binding protein/cell adhesion molecule-like, OBCAMopiate binding-cell adhesion molecule, OPCM, opioid binding protein/cell adhesion molecule-like, opioid binding protein/cell adhesion molecule-like preprotein, Opioid-binding cell adhesion molecule, opioid-binding protein/cell adhesion molecule |
| Host Species | Rabbit |
| Immunogen | The immunogen for Anti-OBCAM/OPCML antibody is: synthetic peptide directed towards the C-terminal of Rat OPCM (NP_446300.1). Peptide sequence EDEYLEISDIKRDQSGEYECSALNDVAAPDVRKVKITVNYPPYISKAKNT |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?