missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Obatoclax Mesylate
GREENER_CHOICE

Product Code. 18729226
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18729226

Brand: Tocris Bioscience™ 6539/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibitor of Bcl-2 family members (IC50 values are 1.11 - 7 μM); antiapoptotic

Obatoclax mesylate is an inhibitor of Bcl-2 family members (IC50 values are 1.11 - 7 μM). Inhibits the interaction of a BH3 domain peptide with 6 members of the Bcl-2 family (Bcl-2, Bcl-XL, Mcl-1, Bcl-W, A1, and Bcl-B) in vitro. Has antiapoptotic and antitumor effects in several tumor bearing mouse models.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[2-[(3,5-Dimethyl-1H-pyrrol-2-yl)methylene)-3-methoxy-2H-pyrrol-5-yl]-1H-indole methanesulfonate
CAS 803712-79-0
Synonym GX15-070
Quantity 10 mg
Molecular Formula C20H19N3O.CH3SO3H
Formula Weight 413.49
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.