missing translation for 'onlineSavingsMsg'
Learn More

OBA-09, MedChemExpress

Product Code. 16485388
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16485388 10 mg 10mg
16448107 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16485388 Supplier Medchem Express Supplier No. HY12840
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress OBA-09, a simple ester of pyruvate and salicylic acid, is potent multi-modal neuroprotectant. OBA-09 has anti-oxidative and anti-inflammatory effects.

OBA-09, a simple ester of pyruvate and salicylic acid, is potent multi-modal neuroprotectant. OBA-09 has anti-oxidative and anti-inflammatory effects.

Specifications

Chemical Name or Material OBA-09
CAS 856095-68-6
Color White
Purity Grade Notes Research
Molecular Formula C10H8O5
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (480.38 mM)
SMILES O=C(O)C1=CC=CC=C1OC(C(C)=O)=O
Molecular Weight (g/mol) 208.17
Formula Weight 208.17
Percent Purity 99.86%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.