missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ OB 24 hydrochloride

Product Code. 18785696
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18785696 10 mg 10mg
18745456 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18785696 Supplier R&D Systems™ Supplier No. 6119/10
  • ONLINE EXCLUSIVE PRICE
    £195.35
    £174.30 / 10mg
    Save £21.05
    10% Off
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective heme oxygenase 1 (HO-1) inhibitor

OB 24 hydrochloride is a potent and selective heme oxygenase 1 (HO-1) inhibitor (IC50 values are 1.9 and >100 μM for HO-1 and HO-2 respectively), reduces protein carbonylation and reactive oxygen species formation in advanced prostate cancer cells (PCA). Inhibits cell proliferation in vitro and PCA tumor growth and lymph node/lung metastases in vivo. Synergizes with Taxol (Cat. No. 1097).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-[[2-[2-(4-Bromophenyl)ethyl]-1,3-dioxolan-2-yl]methyl]-1H-imidazole hydrochloride
CAS 939825-12-4
Assay Percent Range >98%
Quantity 10 mg
Product Line Tocris
Molecular Formula C15H17BrN2O2.HCl
Formula Weight Observed MW: 373.67
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.