missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ OB-1

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
OB-1 is a stomatin-like protein-3 (STOML3) oligomerization inhibitor. Reduces STOML3 cluster size in the plasma membrane. Decreases mechanically-induced Piezo1 but not ASIC3 current amplitudes in N2a cells and cultured mouse sensory neurons in vitro. Subcutaneous administration in mice reversibly silences touch receptor activity and inhibits paw withdrawal reflex in a mouse neuropathic pain model.
Specifications
Specifications
| Chemical Name or Material | Ethyl 5-(1-methyl-4-nitro-1H-imidazol-5-yloxy)-2-phenylbenzofuran-3-carboxylate |
| CAS | 300803-69-4 |
| Quantity | 10 mg |
| Molecular Formula | C21H17N3O6 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction