missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OAZ3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569979
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 86%; Dog: 93%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 93%; Rat: 93%.
The antizyme family is a group of small proteins that play a role in cell growth and division by regulating the biosynthesis of polyamines such as putrescine, spermidine and spermine. AZ3, also known as OAZ3 (Ornithine decarboxylase antizyme 3), is a 187 amino acid protein that belongs to the ODC antizyme family. AZ3 probably plays a key role in spermatogenesis by regulating the intracellular concentration of polyamines in haploid germ cells because it binds to, and destabilizes, ornithine decarboxylase. However, mutations in the AZ3 gene are not a common cause of male infertility and the normal AZ3 protein does not accelerate ornithine decarboxylase degeneration. AZ3 expression has a sharp onset in early spermatids, peaks in later stages and is gone in spermatozoa.
Specifications
| OAZ3 | |
| Polyclonal | |
| Unconjugated | |
| OAZ3 | |
| antizyme 3; AZ3; Az-3; OAZ3; OAZ-t; ODC-Az 3; ornithine decarboxylase antizyme 3; testicular secretory protein Li 31; testis specific ornithine decarboxylase antizyme; TISP15 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 51686 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9UMX2 | |
| OAZ3 | |
| synthetic peptide directed towards the middle region of human OAZ3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction