missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OATP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA526615
This item is not returnable.
View return policy
Description
OATP2 Polyclonal Antibody for Western Blot
A liver-specific member of the organic anion transporter family. This protein is a transmembrane receptor that mediates the sodium-independent uptake of numerous endogenous compounds including bilirubin, 17-beta-glucuronosyl estradiol and leukotriene C4. This protein is also involved in the removal of drug compounds such as statins, bromosulfophthalein and rifampin from the blood into the hepatocytes. Polymorphisms in the gene encoding this protein are associated with impaired transporter function.
Specifications
| OATP2 | |
| Polyclonal | |
| Unconjugated | |
| SLCO1B1 | |
| HBLRR; liver-specific organic anion transporter 1; LST1; LST-1; OATP1B1; OATP2; OATP-2; OATPC; OATP-C; SLC01B1; SLC21A6; SLCO1B1; Sodium-independent organic anion-transporting polypeptide 2; solute carrier family 21 (organic anion transporter), member 6; Solute carrier family 21 member 6; Solute carrier organic anion transporter family member 1B1; solute carrier organic anion transporter family, member 1B1 | |
| Rabbit | |
| Protein A, Antigen affinity chromatography | |
| RUO | |
| 10599 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9Y6L6 | |
| SLCO1B1 | |
| KLH conjugated synthetic peptide between 267-293 amino acids from the central region of human SLCO1B1. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction