missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OATP1B3/SLCO1B3/OATP8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-54739-25ul
This item is not returnable.
View return policy
Description
OATP1B3/SLCO1B3/OATP8 Polyclonal specifically detects OATP1B3/SLCO1B3/OATP8 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| OATP1B3/SLCO1B3/OATP8 | |
| Polyclonal | |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Liver-specific organic anion transporter 2, liver-specific organic anion transporter 3TM13, LST2, LST-2, LST3, OATP1B3LST-3TM13, OATP8, OATP-8, Organic anion transporter 8, organic anion transporter LST-3c, Organic anion-transporting polypeptide 8, SLC21A8, solute carrier family 21 (organic anion transporter), member 8, Solute carrier family 21 member 8, solute carrier organic anion transporter family member 1B3, solute carrier organic anion transporter family, member 1B3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| SLCO1B3 | |
| This antibody was developed against a Recombinant Protein corresponding to amino acids:DVPLSYCNSECNCDESQWEPVCGNNG | |
| 25 μL | |
| Cancer | |
| 28234 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction