missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OATP1B3/SLCO1B3/OATP8 Antibody (CL3770) [HRP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61636H
This item is not returnable.
View return policy
Description
OATP1B3/SLCO1B3/OATP8 Monoclonal antibody specifically detects OATP1B3/SLCO1B3/OATP8 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| OATP1B3/SLCO1B3/OATP8 | |
| Monoclonal | |
| HRP | |
| Liver-specific organic anion transporter 2, liver-specific organic anion transporter 3TM13, LST2, LST-2, LST3, OATP1B3LST-3TM13, OATP8, OATP-8, Organic anion transporter 8, organic anion transporter LST-3c, Organic anion-transporting polypeptide 8, SLC21A8, solute carrier family 21 (organic anion transporter), member 8, Solute carrier family 21 member 8, solute carrier organic anion transporter family member 1B3, solute carrier organic anion transporter family, member 1B3 | |
| This antibody was developed using a recombinant protein derived from Q9NPD5, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer | |
| 28234 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL3770 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction