missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OATP1B1/OATP2 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81750UV
This item is not returnable.
View return policy
Description
OATP1B1/OATP2 Polyclonal antibody specifically detects OATP1B1/OATP2 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| OATP1B1/OATP2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 350 | |
| Liver-specific organic anion transporter 1, LST1SLC21A6, LST-1Solute carrier family 21 member 6, OATP1B1Sodium-independent organic anion-transporting polypeptide 2, OATP2MGC133282, OATP-COATP-2, OATPCsolute carrier organic anion transporter family member 1B1, solute carrier family 21 (organic anion transporter), member 6, solute carrier organic anion transporter family, member 1B1 | |
| OATP1B1 antibody was raised against a 14 amino acid peptide near the carboxy terminus of human OATP1B1. The immunogen is located within the last 50 amino acids of OATP1B1. Amino Acid Squence: MKKKYQEKDINASE | |
| 0.1 mL | |
| Membrane Trafficking and Chaperones | |
| 10599 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction