missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OAT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106699
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total OAT.
OAT is the mitochondrial enzyme ornithine aminotransferase, which is a key enzyme in the pathway that converts arginine and ornithine into the major excitatory and inhibitory neurotransmitters glutamate and GABA.
Specifications
| OAT | |
| Polyclonal | |
| Unconjugated | |
| OAT | |
| AI194874; GACR; gyrate atrophy; HOGA; OAT; OATASE; OKT; ornithine aminotransferase; ornithine aminotransferase (gyrate atrophy); Ornithine aminotransferase, hepatic form; Ornithine aminotransferase, mitochondrial; Ornithine aminotransferase, renal form; Ornithine delta-aminotransferase; ornithine-delta-aminotransferase; ornithine--oxo-acid aminotransferase; ornithine-oxo-acid aminotransferase; rOAT; testicular tissue protein Li 130 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18242, 4942, 64313 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P04181, P04182, P29758 | |
| OAT | |
| A synthesized peptide derived from human OAT(Accession P04181), corresponding to amino acid residues T128-W178. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction