Learn More
Invitrogen™ OAT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527968
Description
Recommended positive controls: K562, THP-1, Mouse brain, OAT-transfected 293T. Predicted reactivity: Mouse (93%), Rat (93%), Zebrafish (81%), Xenopus laevis (84%), Pig (94%), Chicken (86%), Rhesus Monkey (97%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
OAT is the mitochondrial enzyme ornithine aminotransferase, which is a key enzyme in the pathway that converts arginine and ornithine into the major excitatory and inhibitory neurotransmitters glutamate and GABA.
Specifications
| OAT | |
| Polyclonal | |
| Unconjugated | |
| OAT | |
| AI194874; GACR; gyrate atrophy; HOGA; OAT; OATASE; OKT; ornithine aminotransferase; ornithine aminotransferase (gyrate atrophy); Ornithine aminotransferase, hepatic form; Ornithine aminotransferase, mitochondrial; Ornithine aminotransferase, renal form; Ornithine delta-aminotransferase; ornithine-delta-aminotransferase; ornithine--oxo-acid aminotransferase; ornithine-oxo-acid aminotransferase; rOAT; testicular tissue protein Li 130 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18242, 4942 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.28 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P04181, P29758 | |
| OAT | |
| Recombinant protein encompassing a sequence within the center region of human OAT. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.