missing translation for 'onlineSavingsMsg'
Learn More

OASL Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18425765 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18425765 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18425765 Supplier Novus Biologicals Supplier No. NBP281833AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

OASL Polyclonal antibody specifically detects OASL in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen OASL
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias 2'-5'-oligoadenylate synthetase-like, p59 OASL, p59OASL59 kDa 2'-5'-oligoadenylate synthase-like protein, Thyroid receptor-interacting protein 14,59 kDa 2'-5'-oligoadenylate synthetase-like protein, TRIP-14, TRIP14TR-interacting protein 14
Host Species Rabbit
Immunogen OASL antibody was raised against a 17 amino acid peptide near the amino terminus of human OASL. The immunogen is located within the first 50 amino acids of OASL. Amino Acid Squence: VAQWLQPHREWKEEVLD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 8638
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.