missing translation for 'onlineSavingsMsg'
Learn More

OASIS/CREB3L1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody

Brand:  Novus Biologicals NB600-799

Product Code. 18037263

  • £399.00 / 0.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

OASIS/CREB3L1 Polyclonal specifically detects OASIS/CREB3L1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

OASIS/CREB3L1
Polyclonal
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
BBF-2 homolog, cAMP responsive element binding protein 3-like 1, cAMP-responsive element-binding protein 3-like protein 1, OASIScyclic AMP-responsive element-binding protein 3-like protein 1, old astrocyte specifically induced substance, Old astrocyte specifically-induced substance
Goat
Affinity Purified
RUO
90993
Human
IgG
Western Blot, Immunohistochemistry (Paraffin), ELISA
0.5 mg/mL
Western Blot 1 - 3 ug/ml, Immunohistochemistry-Paraffin 3.75 ug/ml, Peptide ELISA Detection limit 1:32000
NP_443086.1
CREB3L1
Peptide with sequence C-DFSHSKEWFHDRD corresponding to C-Terminus according to NP_443086.1.
0.1 mg
Primary
Human. Predicted cross-reactivity based on sequence identity: Mouse and Rat.
Store at -20C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only