missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OAS2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565422
This item is not returnable.
View return policy
Description
Immunogen sequence: TLVLFFSDLKQ FQDQKRSQRD ILDKTGDKLK FCLFTKWLKN NFEIQKSLDG FTIQVFTKNQ RISFEVLAAF NALSLNDNPS PWIYRELKRS LDKTNASPGE FAVCFTELQQ K Highest antigen sequence identity to the following orthologs - mouse 42%, rat 44%.
This gene encodes a member of the 2-5A synthetase family, essential proteins involved in the innate immune response to viral infection. The encoded protein is induced by interferons and uses adenosine triphosphate in 2'-specific nucleotidyl transfer reactions to synthesize 2',5'-oligoadenylates. These molecules activate latent RNase L, which results in viral RNA degradation and the inhibition of viral replication. The three known members of this gene family are located in a cluster on chromosome 12. Alternatively spliced transcript variants encoding different isoforms have been described.
Specifications
| OAS2 | |
| Polyclonal | |
| Unconjugated | |
| OAS2 | |
| (2-5')oligo(A) synthase 2; (2'-5')oligo(A) synthetase 2; 2',5'-oligoadenylate synthetase-like 11; 2,5-oligoadenylate synthetase-like 11; 2'-5' oligoadenylate synthetase 2; 2'-5' oligoadenylate synthetase-like 11; 2-5A synthase 2; 2'-5'-oligoadenylate synthase 2; 2'-5'-oligoadenylate synthetase 2; 2'-5'-oligoadenylate synthetase 2, 69/71kDa; A synthetase 2; MGC78578; Oas2; Oasl11; p69 OAS / p71 OAS; p69OAS / p71OAS | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4939 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P29728 | |
| OAS2 | |
| Recombinant Human OAS2. Recombinant protein control fragment (Product #RP-106105). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction