missing translation for 'onlineSavingsMsg'
Learn More

OAS2 Antibody (OTI6E6), FITC, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-73137F

Product Code. 30039596

  • £587.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

OAS2 Monoclonal antibody specifically detects OAS2 in Human samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

OAS2
Monoclonal
FITC
PBS
Mouse
Immunogen affinity purified
RUO
4939
Store at 4C in the dark.
IgG2a
Western Blot, Immunohistochemistry
OTI6E6
Western Blot, Immunohistochemistry
(2-5')oligo(A) synthase 2, (2'-5')oligo(A) synthetase 2, (2-5')oligo(A) synthetase 2, 2'5' oligoadenylate synthetase 2, 2-5A synthase 2, 2-5A synthetase 2, 2'-5'-oligoadenylate synthase 2, 2'-5'-oligoadenylate synthetase 2 (69-71 kD), 2'-5'-oligoadenylate synthetase 2, 69/71kDa, EC 2.7.7, EC 2.7.7.-, MGC78578, p69 OAS/ p71 OAS, p69OAS/ p71OAS
Human recombinant protein fragment corresponding to amino acids 60-335 of human OAS2 (NP_002526) produced in E.coli.
0.1 mL
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.