missing translation for 'onlineSavingsMsg'
Learn More

Cole Parmer Oakton pHTestr™ 30 Waterproof Pocket Tester with Calibration Kit

Product Code. 16627105
Click to view available options
Calibration Points:
3
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 16627105

Brand: Cole Parmer 3563490

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Oakton pHTestr™ 30 Waterproof Pocket Tester with Calibration Kit features a versatile, compact design that works in a field or laboratory setting.

Waterproof pHTestr 30 is great for outdoor applications such as taking pH measurements in lakes and streams. Designed to float in water, you can even attach a line to the housing for easy retrieval. Valox sensor housing resists most chemicals.

  • Ideal for outdoor applications such as taking pH measurements in lakes and streams.
  • Valox sensor housing resists most chemicals
  • 0.01 pH accuracy along with dual pH and selectable C or F temperature display
  • Replaceable, double-junction electrode sensors
  • IP67-rated housing sits flat on the table and floats in water
  • Replaceable sensor for easy maintenance.
  • Double-junction electrode sensor handles wastewater and solutions containing sulfides, heavy metals, and TRIS.
  • Electrode sensor offers a clog-resistant Kynar™ junction and a recessed, glass pH bulb to protect against breakage.
  • Keypad clicks to confirm operation, and allows access to a variety of functions.
  • Calibrate up to three pH points with the auto-buffer recognition at both USA and NIST standard buffers.
  • Additional features include a automatic temperature compensation (ATC), hold function, auto-off function, self-diagnostic error messages, and a three-segment battery-life indicator.
  • Meter kit adds: two single-use pouches of pH 4.01, pH 7.00, pH 10.0 buffer, and rinse solution, one sample jar, and sturdy carrying case.
  • LCD Display
  • pH range: 0 to 14
  • pH Resolution: 0.01/0.1
  • pH Accuracy: ±0.01 pH
  • Buffer Recognition: NIST and USA
  • Temp range: -5 to 50°C (23 to 122°F)
  • Automatic temperature compensation
  • Waterproof
  • Includes 4 X A76 batteries
  • Diameter: 1.5 in. (3.81 cm)

Specifications

Calibration Up to 3 points
Calibration Points 3
Type Pocket Tester
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.