missing translation for 'onlineSavingsMsg'
Learn More

pH Calibration Buffer Bottles, Oakton™

Product Code. 10303765 Shop All Cole-Parmer Products
Change view
Click to view available options
pH:
1.68
7.00
10.00
12.46
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH unitSize
10303765 7.00 Each
11739679 1.68 Each
11750079 10.00 Each
11710069 12.46 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10303765 Supplier Cole Parmer Supplier No. 0065404

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use these PH buffers for meters with 0.01 resolution. Oakton™ pH Calibration Buffer Bottles are standardized against NIST-traceable pH references within 0.01 pH at 77°F (25°C).

  • pH buffer solutions allow the user to adjust their system to read accurate measurements
  • Store buffers away from direct sunlight.

Order Info

Shipping weight: 1-1/4 lbs. (0.6kg).

Certifications

NIST traceable, Certified to NIST traceable standards.

Specifications

For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 7.00
Physical Form Liquid
Quantity 500 mL
Solution Type pH Buffer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.