missing translation for 'onlineSavingsMsg'
Learn More

OAC2, MedChemExpress

Product Code. 16486026
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486026 100 mg 100mg
16431617 50 mg 50mg
16425438 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16486026 Supplier Medchem Express Supplier No. HY12884

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress OAC2 is an Oct4-activating compound which activates expression through the Oct4 gene promoter.

OAC2 is an Oct4-activating compound which activates expression through the Oct4 gene promoter.

Specifications

Chemical Name or Material OAC2
Physical Form Solid
CAS 6019-39-2
Color Gray
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C15H12N2O
Solubility Information DMSO : ≥ 100 mg/mL (423.24 mM) ∣Ethanol : 25 mg/mL (105.81 mM; Need ultrasonic)
SMILES O=C(NC1=CC2=C(NC=C2)C=C1)C3=CC=CC=C3
Molecular Weight (g/mol) 236.27
Formula Weight 236.27
Percent Purity 99.57%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.