missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ OAC-2
Oct4 activator; enhances iPSC reprogramming efficiency
Specifications
| Chemical Name or Material | OAC-2 |
|---|---|
| Physical Form | Solid |
| Product Type | Bioactive Small Molecule |
| Solubility | Soluble to 100mM in DMSO and to 100mM in ethanol |
| Synonym | N-(1H-Indol-5-yl)benzamide |
Description
Oct 4 activator; enhances iPSC reprogramming four-fold and accelerates the appearance of iPSC colonies in combination with 4F (Oct4, Sox2, c-Myc and Klf4). Also induces cardiomyocyte-like cells from human fibroblasts, as part of the 9C cocktail ( CHIR99021, A83-01, BIX01294, AS83541, SC1, Y27632, SU 16f, JNJ10198409 and OAC-2)Specifications
| OAC-2 | |
| Bioactive Small Molecule | |
| N-(1H-Indol-5-yl)benzamide |
| Solid | |
| Soluble to 100mM in DMSO and to 100mM in ethanol | |
| 6019-39-2 |
Safety and Handling
Recommended Storage : Store at 4°C
For Research Use Only.
Spot an opportunity for improvement?
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.