missing translation for 'onlineSavingsMsg'
Learn More

O6-Benzylguanine, 98%

Product Code. 15496748
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 15496748

Brand: Thermo Scientific Alfa Aesar H60274.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19916-73-5 | C12H11N5O | 241.25 g/mol

O6-Benzylguanine is an effective substrate and efficient inactivator of DNA alkyltransferase. O6-Benzylguanine is an inhibitor of MGMT. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O6-Benzylguanine is an effective substrate and efficient inactivator of DNA alkyltransferase. O6-Benzylguanine is an inhibitor of MGMT. It is also used as an important raw material and intermediate used in organic Synthesis, pharmac
TRUSTED_SUSTAINABILITY

Specifications

CAS 19916-73-5
Molecular Formula C12H11N5O
MDL Number MFCD00269931
Quantity 250 mg
Synonym o6-benzylguanine, 6-o-benzylguanine, 6-benzyloxy-7h-purin-2-amine, o 6-benzylguanine, 2-amino-6-benzyloxypurine, 6-benzyloxy-9h-purin-2-amine, o 6-bgua, 6-benzyloxy guanine, 2-amino-6-benzyloxy purine, 6-benzyloxyguanine
Solubility Information Soluble in methanol (20mg/ml),and aqueous alkali (Heated).
InChI Key KRWMERLEINMZFT-UHFFFAOYSA-N
SMILES NC1=NC(OCC2=CC=CC=C2)=C2NC=NC2=N1
IUPAC Name 6-phenylmethoxy-7H-purin-2-amine
Molecular Weight (g/mol) 241.25
PubChem CID 4578
Formula Weight 241.25
Percent Purity 98%
Chemical Name or Material O6-Benzylguanine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.