missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ O4I2
GREENER_CHOICE

Product Code. 18798208
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18798208

Brand: Tocris Bioscience™ 5664/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Oct3/4 inducer; induces expression of pluripotent-associated genes

O4I2 is an Oct3/4 inducer; promotes Oct3/4 expression and stabilizes the Oct3/4 protein in HEK293 cells. Induces expression of pluripotent-associated genes Lin28, Sox2 and Nanog, and suppresses Rex1. Induces 15-fold increase in Oct3/4 expression in human primary fibroblasts.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl 2-[(4-chlorophenyl)amino]-4-thiazolecarboxylate
CAS 165682-93-9
Quantity 10 mg
Target Additional Stem Cell Dedifferentiation Products
Molecular Formula C12H11ClN2O2S
Formula Weight 282.75
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.