missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ O4I2

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
O4I2 is an Oct3/4 inducer; promotes Oct3/4 expression and stabilizes the Oct3/4 protein in HEK293 cells. Induces expression of pluripotent-associated genes Lin28, Sox2 and Nanog, and suppresses Rex1. Induces 15-fold increase in Oct3/4 expression in human primary fibroblasts.
Specifications
Specifications
| Chemical Name or Material | Ethyl 2-[(4-chlorophenyl)amino]-4-thiazolecarboxylate |
| CAS | 165682-93-9 |
| Quantity | 50 mg |
| Target | Additional Stem Cell Dedifferentiation Products |
| Molecular Formula | C12H11ClN2O2S |
| Formula Weight | 282.75 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction