missing translation for 'onlineSavingsMsg'
Learn More

O4I1, MedChemExpress

Product Code. 16493637
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493637 50 mg 50mg
16477628 10 mg 10mg
16467536 100 mg 100mg
16431286 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16493637 Supplier Medchem Express Supplier No. HY18771
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress O4I1 is as a potent Oct3/4 inducer.

O4I1 is as a potent Oct3/4 inducer.

Specifications

Chemical Name or Material O4I1
CAS 175135-47-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C16H15NO2
Quantity 50 mg
Solubility Information DMSO : ≥ 38 mg/mL (150.02 mM)
SMILES N#CCC1=CC=C(OCC2=CC=C(OC)C=C2)C=C1
Molecular Weight (g/mol) 253.3
Formula Weight 253.3
Percent Purity 95.3%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.