missing translation for 'onlineSavingsMsg'
Learn More

O4-Ethylthymidine, TRC

Product Code. 30384673
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384673 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384673 Supplier Toronto Research Chemicals Supplier No. TRCE677520250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material O4-Ethylthymidine
CAS 59495-22-6
Molecular Formula C12 H18 N2 O5
Quantity 250 mg
Synonym 4-O-Ethyl-thymidine, 4-O-Ethylthymidine, 1-(2-Deoxy-β-D-erythro-pentofuranosyl)-4-ethoxy-5-methyl-2(1H)-pyrimidinone, O4-Ethylthymidine, 4-Ethoxy-1-(2R-4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidin-2(1H)-one
InChI Formula InChI=1S/C12H18N2O5/c1-3-18-11-7(2)5-14(12(17)13-11)10-4-8(16)9(6-15)19-10/h5,8-10,15-16H,3-4,6H2,1-2H3/t8-,9+,10+/m0/s1
SMILES CCOC1=NC(=O)N(C=C1C)[C@H]2C[C@H](O)[C@@H](CO)O2
IUPAC Name 4-ethoxy-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidin-2-one
Molecular Weight (g/mol) 270.282
Formula Weight 270.122
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.