missing translation for 'onlineSavingsMsg'
Learn More

o-Xylylene dichloride, 98%

Product Code. p-7020516
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11484134

Brand: Thermo Scientific Alfa Aesar A12500.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 612-12-4 | C8H8Cl2 | 175.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 612-12-4
Molecular Formula C8H8Cl2
Molecular Weight (g/mol) 175.05
MDL Number MFCD00000903
InChI Key FMGGHNGKHRCJLL-UHFFFAOYSA-N
Synonym 1,2-bis chloromethyl benzene, o-xylylene dichloride, alpha,alpha'-dichloro-o-xylene, o-bis chloromethyl benzene, benzene, 1,2-bis chloromethyl, ccris 1773, o-xylene, alpha,alpha'-dichloro, benzene, 1,2-dichloromethyl, .alpha.,.alpha.'-dichloro-o-xylene, o-xylylenchlorid
PubChem CID 11919
IUPAC Name 1,2-bis(chloromethyl)benzene
SMILES ClCC1=CC=CC=C1CCl

Specifications

Melting Point 53°C to 57°C
Boiling Point 246°C
Flash Point 107°C (224°F)
Odor Pungent
Quantity 50 g
UN Number UN2928
Beilstein 2043675
Formula Weight 175.06
Percent Purity 98%
Chemical Name or Material o-Xylylene dichloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.