missing translation for 'onlineSavingsMsg'
Learn More

o-Xylene, TRC

Product Code. 30456168
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456168 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 30456168 Supplier Toronto Research Chemicals Supplier No. TRCX749405500ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material o-Xylene (1,2-Dimethylbenzene)
CAS 95-47-6
Molecular Formula C8H10
Quantity 500 mL
Synonym Benzene','1,2-dimethyl-, o-Xylene (8CI), 1,2-Dimethylbenzene, 1,2-Xylene, 2-Methyltoluene, 3,4-Dimethylbenzene, NSC 60920, Orthoxylene, o-Dimethylbenzene, o-Methyltoluene, o-Xylol, ortho-Xylene
InChI Formula InChI=1S/C8H10/c1-7-5-3-4-6-8(7)2/h3-6H,1-2H3
SMILES Cc1ccccc1C
IUPAC Name 1,2-xylene
Molecular Weight (g/mol) 106.16
Formula Weight 106.08
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.