missing translation for 'onlineSavingsMsg'
Learn More

o-Xylene, 99%

Product Code. 11410163
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410163 2500 mL 2500mL
11468700 100 mL 100mL
11400163 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 11410163 Supplier Thermo Scientific Alfa Aesar Supplier No. A11358.0F
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 95-47-6 | C8H10 | 106.17 g/mol

o-Xylene is widely used in the production of phthalic anhydride. It is also used in fuels as well as fuel additives. It can be used in the production of isophthalic acid and terephthalic acid. It is utilized in polyvinyl chloride production, alkyl resin and unsaturated polyester resins. It is also used as a solvent in paint, varnish, glue, printing ink, dyes and in rubber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Xylene is widely used in the production of phthalic anhydride. It is also used in fuels as well as fuel additives. It can be used in the production of isophthalic acid and terephthalic acid. It is utilized in polyvinyl chloride production, alkyl resin and unsaturated polyester resins. It is also used as a solvent in paint, varnish, glue, printing ink, dyes and in rubber.

Solubility
Miscible with ethanol, diethylether. Immiscible with water.

Notes
Incompatible with oxidizing agents and strong acids
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-47-6
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008519
InChI Key CTQNGGLPUBDAKN-UHFFFAOYSA-N
Synonym o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes
PubChem CID 7237
ChEBI CHEBI:28063
IUPAC Name 1,2-xylene
SMILES CC1=CC=CC=C1C

Specifications

Melting Point -25°C
Density 0.877
Quantity 2500 mL
Flash Point 32°C (89°F)
Refractive Index 1.505
Linear Formula (CH3)2C6H4
UN Number UN1307
Beilstein 1815558
Merck Index 14,10081
Boiling Point 143°C to 145°C
Solubility Information Miscible with ethanol,diethylether. Immiscible with water.
Formula Weight 106.17
Percent Purity 99%
Chemical Name or Material o-Xylene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • o-Xylene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: o-Xylene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.