Learn More
o-Xylene, 99%
Description
o-Xylene is widely used in the production of phthalic anhydride. It is also used in fuels as well as fuel additives. It can be used in the production of isophthalic acid and terephthalic acid. It is utilized in polyvinyl chloride production, alkyl resin and unsaturated polyester resins. It is also used as a solvent in paint, varnish, glue, printing ink, dyes and in rubber.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
o-Xylene is widely used in the production of phthalic anhydride. It is also used in fuels as well as fuel additives. It can be used in the production of isophthalic acid and terephthalic acid. It is utilized in polyvinyl chloride production, alkyl resin and unsaturated polyester resins. It is also used as a solvent in paint, varnish, glue, printing ink, dyes and in rubber.
Solubility
Miscible with ethanol, diethylether. Immiscible with water.
Notes
Incompatible with oxidizing agents and strong acids
Specifications
Specifications
| Melting Point | -25°C |
| Density | 0.877 |
| Boiling Point | 143°C to 145°C |
| Quantity | 100 mL |
| Flash Point | 32°C (89°F) |
| Refractive Index | 1.505 |
| Linear Formula | (CH3)2C6H4 |
| UN Number | UN1307 |
| Beilstein | 1815558 |
| Merck Index | 14,10081 |
| Show More |
Sicherheit und Handhabung
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : o-Xylene
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.