missing translation for 'onlineSavingsMsg'
Learn More

O-Torsemide, TRC

Product Code. 30450654
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30450654 500 mg 500mg
30450326 250 mg -
30449146 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30450654 Supplier Toronto Research Chemicals Supplier No. TRCT585500500MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106944-62-1
Molecular Formula C16 H20 N4 O3 S
Molecular Weight (g/mol) 348.42
Synonym 3-Pyridinesulfonamide, N-[[(1-methylethyl)amino]carbonyl]-4-[(2-methylphenyl)amino]-, o-Torasemide, 1-(1-Methylethyl)-3-[[4-[(2-methylphenyl)amino]pyridin-3-yl]sulfonyl]urea
IUPAC Name 1-[4-(2-methylanilino)pyridin-3-yl]sulfonyl-3-propan-2-ylurea
SMILES CC(C)NC(=O)NS(=O)(=O)c1cnccc1Nc2ccccc2C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C16H20N4O3S/c1-11(2)18-16(21)20-24(22,23)15-10-17-9-8-14(15)19-13-7-5-4-6-12(13)3/h4-11H,1-3H3,(H,17,19)(H2,18,20,21)
Formula Weight 348.1256
Chemical Name or Material o-Torasemide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.