missing translation for 'onlineSavingsMsg'
Learn More

o-Tolylmagnesium chloride, 1.4M (22 wt.%) solution in THF/toluene, AcroSeal™, Thermo Scientific Chemicals

Product Code. p-3771093 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10103494 100 mL AcroSeal™ Glass bottle 100mL
10682255 800 mL AcroSeal™ Glass bottle 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10103494 Supplier Thermo Scientific Acros Supplier No. 427481000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33872-80-9 , 109-99-9 , 108-88-3
Molecular Formula C7H7ClMg
Molecular Weight (g/mol) 150.89
MDL Number MFCD00010355
InChI Key LQVSLLSEXLZRRH-UHFFFAOYSA-M
Synonym o-tolylmagnesium chloride, magnesium, chloro 2-methylphenyl, o-tolylmagnesium chloride solution, 2-methylphenylmagnesium chloride, unii-gul3abu3p5, o-tolyl magnesium chloride, gul3abu3p5, chloro-o-tolylmagnesium, o-tolylmagnesiumchloride, 2-tolyl magnesium chloride
PubChem CID 118563
IUPAC Name magnesium;methylbenzene;chloride
SMILES [Mg++].[Cl-].Cc1cccc[cH-]1

Specifications

Density 0.9600g/mL
Flash Point −21°C
Packaging AcroSeal™ Glass bottle
Linear Formula CH3C6H4MgCl
Quantity 100 mL
Specific Gravity 0.96
Formula Weight 150.89
Chemical Name or Material o-Tolylmagnesium chloride, 1.4M (22 wt.%) solution in THF/toluene
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • o-Tolylmagnesium chloride, 1.4M (22 wt.%) solution in THF/touene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.