missing translation for 'onlineSavingsMsg'
Learn More

o-Tolylhydrazine Hydrochloride, TRC

Product Code. 30450250
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30450250 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30450250 Supplier Toronto Research Chemicals Supplier No. TRCT536740100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material o-Tolylhydrazine Hydrochloride
CAS 635-26-7
Molecular Formula C7H11ClN2
Quantity 100 g
Synonym (2-Methylphenyl)hydrazine Monohydrochloride, o-Tolyl-Hydrazine Monohydrochloride, 2-Methylbenzenehydrazine Hydrochloride2-Methylphenylhydrazine Hydrochloride
InChI Formula InChI=1S/C7H10N2.ClH/c1-6-4-2-3-5-7(6)9-8;/h2-5,9H,8H2,1H3;1H
SMILES Cl.Cc1ccccc1NN
IUPAC Name (2-methylphenyl)hydrazine;hydrochloride
Molecular Weight (g/mol) 158.63
Formula Weight 158.0611
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.