missing translation for 'onlineSavingsMsg'
Learn More

o-Tolylhydrazine hydrochloride, 98%

Product Code. 11432428
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11432428 5 g 5g
11442428 25 g 25g
11452428 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11432428 Supplier Thermo Scientific Alfa Aesar Supplier No. A15767.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 635-26-7 | C7H11ClN2 | 158.629 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 635-26-7
Molecular Formula C7H11ClN2
Molecular Weight (g/mol) 158.629
MDL Number MFCD00012925
InChI Key KJGFNDCSTWGUDT-UHFFFAOYSA-N
Synonym o-tolylhydrazine hydrochloride, 2-methylphenyl hydrazine hydrochloride, 2-methylphenylhydrazine hydrochloride, o-tolylhydrazine hcl, 1-2-methylphenyl hydrazine hydrochloride, 2-tolylhydrazine hydrochloride, o-tolylhydrazinium 1+ chloride, o-methylphenylhydrazine hcl, 2-methylphenylhydrazine hcl, hydrazine, 2-methylphenyl-, monohydrochloride
PubChem CID 71554
IUPAC Name (2-methylphenyl)hydrazine;hydrochloride
SMILES CC1=CC=CC=C1NN.Cl

Specifications

Melting Point ∼190°C (decomposition)
Quantity 5 g
UN Number UN2811
Beilstein 3655929
Sensitivity Hygroscopic
Formula Weight 158.63
Percent Purity 98%
Chemical Name or Material o-Tolylhydrazine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.