missing translation for 'onlineSavingsMsg'
Learn More

o-Toluoyl chloride, 99%

Product Code. 10438500 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10438500 100 g Glass bottle 100g
10552551 5 g Glass bottle 5g
10357230 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10438500 Supplier Thermo Scientific Acros Supplier No. 153371000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 933-88-0 | C8H7ClO | 154.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 933-88-0
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.59
MDL Number MFCD00000668
InChI Key GPZXFICWCMCQPF-UHFFFAOYSA-N
Synonym o-toluoyl chloride, benzoyl chloride, 2-methyl, o-toluoylchloride, methylbenzoyl chloride, o-methylbenzoyl chloride, 2-toluoyl chloride, o-toluic acid chloride, unii-j9ykn18e7r, o-toluyl chloride, 2-methylbenzoic acid chloride
PubChem CID 70276
IUPAC Name 2-methylbenzoyl chloride
SMILES CC1=CC=CC=C1C(Cl)=O

Specifications

Color Pink to Yellow
Density 1.1800g/mL
Boiling Point 88°C to 90°C (12.0 mmHg)
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3C6H4COCl
Refractive Index 1.5540 to 1.556
Quantity 100 g
Beilstein 09, 464
Specific Gravity 1.18
Solubility Information Solubility in water: decomposes
Formula Weight 154.6
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material o-Toluoyl chloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • o-Toluoyl chloride
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.