missing translation for 'onlineSavingsMsg'
Learn More

o-Tolunitrile, 98%

Product Code. 10377850 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
25mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10377850 25 mL Glass bottle 25mL
10143440 250 mL Glass bottle 250mL
10771915 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10377850 Supplier Thermo Scientific Acros Supplier No. 152860250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 529-19-1 | C8H7N | 117.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 529-19-1
Molecular Formula C8H7N
Molecular Weight (g/mol) 117.15
MDL Number MFCD00001792
InChI Key NWPNXBQSRGKSJB-UHFFFAOYSA-N
Synonym o-tolunitrile, 2-cyanotoluene, o-toluonitrile, benzonitrile, 2-methyl, o-cyanotoluene, o-tolylnitrile, o-toluic nitrile, 2-methylbenzenecarbonitrile, o-methylbenzonitrile, 2-tolunitrile
PubChem CID 10721
IUPAC Name 2-methylbenzonitrile
SMILES CC1=CC=CC=C1C#N

Specifications

Melting Point -13.0°C
Color Colorless to Yellow
Density 0.9900g/mL
Boiling Point 205.0°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4CN
Refractive Index 1.5269 to 1.5289
Quantity 25 mL
Beilstein 09, 466
Merck Index 15, 9697
Specific Gravity 0.99
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol and ether
Viscosity 1.7 mm2/s (23°C)
Formula Weight 117.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material o-Tolunitrile, 98%
Show More Show Less
compliance-icons
Product Identifier
  • o-Tolunitrile
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.