missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Toluic-D7 Acid, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | o-Toluic-D7 Acid |
| CAS | 207742-73-2 |
| Molecular Formula | C8 D7 H O2 |
| Quantity | 1 g |
| Synonym | 6-(Methyl-D3)-benzoic-2,3,4,5-D4 Acid, Benzoic-2,3,4,5-d4 acid','6-(methyl-d3)- (9CI','ACI), 6-(Methyl-d3)benzoic-2,3,4,5-d4 acid (ACI), 2,3,4,5-Tetradeuterio-6-(trideuteriomethyl)benzoic acid, 2-Methylbenzoic acid D7 |
| InChI Formula | InChI=1S/C8H8O2/c1-6-4-2-3-5-7(6)8(9)10/h2-5H,1H3,(H,9,10)/i1D3,2D,3D,4D,5D |
| SMILES | [2H]c1c([2H])c([2H])c(c(C(=O)O)c1[2H])C([2H])([2H])[2H] |
| IUPAC Name | 2,3,4,5-tetradeuterio-6-(trideuteriomethyl)benzoic acid |
| Molecular Weight (g/mol) | 143.19 |
| Formula Weight | 143.0964 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction