missing translation for 'onlineSavingsMsg'
Learn More

o-Toluic Acid, TRC

Product Code. 30452086
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30452086 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30452086 Supplier Toronto Research Chemicals Supplier No. TRCT5361205G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 118-90-1
Molecular Formula C8 H8 O2
Quantity 5 g
Synonym Benzoic acid, 2-methyl- (9CI, ACI), 2-Methylbenzoic acid (ACI), o-Toluic acid (8CI), NSC 2193, o-Methylbenzoic acid, o-Toluylic acid
InChI Formula InChI=1S/C8H8O2/c1-6-4-2-3-5-7(6)8(9)10/h2-5H,1H3,(H,9,10)
SMILES Cc1ccccc1C(=O)O
IUPAC Name 2-methylbenzoic acid
Molecular Weight (g/mol) 136.15
Formula Weight 136.0524
Chemical Name or Material o-Toluic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.