missing translation for 'onlineSavingsMsg'
Learn More

o-Toluic acid, 98+%

Product Code. 11439060
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11439060 250 g 250g
11419060 1000 g 1000g
11429060 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439060 Supplier Thermo Scientific Alfa Aesar Supplier No. A13856.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 118-90-1 | C8H8O2 | 136.15 g/mol

o-Toluic acid is used for producing of fluorescent whitening agent ER33%, bensulfuronmethyl,o-tolunitrile,o-cyano-benzaldehyde and 3,5-dinitro-o-toluylamide etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Toluic acid is used for producing of fluorescent whitening agent ER33%, bensulfuronmethyl,o-tolunitrile,o-cyano-benzaldehyde and 3,5-dinitro-o-toluylamide etc.

Solubility
Soluble in alcohol and chloroform. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-90-1
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00002477
InChI Key ZWLPBLYKEWSWPD-UHFFFAOYSA-N
Synonym o-toluic acid, o-toluylic acid, orthotoluic acid, 2-toluic acid, toluic acid, benzoic acid, 2-methyl, o-methylbenzoic acid, 2-methyl-benzoic acid, benzoic acid, methyl, o-toluate
PubChem CID 8373
ChEBI CHEBI:36632
IUPAC Name 2-methylbenzoic acid
SMILES CC1=CC=CC=C1C(=O)O

Specifications

Melting Point 103°C to 106°C
Density 1.06
Boiling Point 258°C to 259°C
Flash Point 148°C (298°F)
Odor Aromatic
Quantity 250 g
Beilstein 1072103
Merck Index 14,9535
Solubility Information Soluble in alcohol and chloroform. Slightly soluble in water.
Formula Weight 136.15
Percent Purity ≥98%
Chemical Name or Material o-Toluic acid
Show More Show Less
compliance-icons
Product Identifier
  • o-Toluic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: o-Toluic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.