missing translation for 'onlineSavingsMsg'
Learn More

o-Toluic acid, 98+%

Product Code. 10224810 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
1 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
1kg
5g
This item is not returnable. View return policy

Product Code. 10224810

Brand: Thermo Scientific Acros 139041000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 118-90-1 | C8H8O2 | 136.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-90-1
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00002477
InChI Key ZWLPBLYKEWSWPD-UHFFFAOYSA-N
Synonym o-toluic acid, o-toluylic acid, orthotoluic acid, 2-toluic acid, toluic acid, benzoic acid, 2-methyl, o-methylbenzoic acid, 2-methyl-benzoic acid, benzoic acid, methyl, o-toluate
PubChem CID 8373
ChEBI CHEBI:36632
IUPAC Name 2-methylbenzoic acid
SMILES CC1=CC=CC=C1C(=O)O

Specifications

Melting Point 103°C to 105°C
Color White to Yellow
Density 1.06
Boiling Point 258°C to 259°C
Flash Point 148°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Plastic bottle
Linear Formula CH3C6H4CO2H
Quantity 100 g
Beilstein 09, 462
Merck Index 14, 9535
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and chloroform
Formula Weight 136.15
Percent Purity 98+%
Physical Form Needle-like Crystalline Solid
Chemical Name or Material o-Toluic acid, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • o-Toluic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.