missing translation for 'onlineSavingsMsg'
Learn More

o-Toluenethiol, 97%

Product Code. 10139499 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10139499 25 g Glass bottle 25g
10310644 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10139499 Supplier Thermo Scientific Acros Supplier No. 259240250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 137-06-4 | C7H8S | 124.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-06-4
Molecular Formula C7H8S
Molecular Weight (g/mol) 124.20
MDL Number MFCD00004838
InChI Key LXUNZSDDXMPKLP-UHFFFAOYSA-N
Synonym o-toluenethiol, o-thiocresol, 2-methylthiophenol, 2-thiocresol, 2-toluenethiol, o-tolyl mercaptan, o-mercaptotoluene, o-methylthiophenol, o-methylbenzenethiol, benzenethiol, 2-methyl
PubChem CID 8712
IUPAC Name 2-methylbenzenethiol
SMILES CC1=CC=CC=C1S

Specifications

Melting Point 14.0°C
Color Colorless to Yellow
Density 1.0540g/mL
Boiling Point 194.0°C
Flash Point 63°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4SH
Refractive Index 1.5790 to 1.5810
Quantity 25 g
Beilstein 06,37
Merck Index 15,9478
Specific Gravity 1.054
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 124.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material o-Toluenethiol, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • o-Toluenethiol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.