missing translation for 'onlineSavingsMsg'
Learn More

o-Tolualdehyde, 98%, stab. with 0.1% hydroquinone

Product Code. 11429462
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429462 500 g 500g
11409462 25 g 25g
11419462 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429462 Supplier Thermo Scientific Alfa Aesar Supplier No. A10172.36

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 529-20-4 | C8H8O | 120.151 g/mol

o-tolualdehyde was used in determination of alkenal-2,4-dinitrophenylhydrazones by HPLC by addition of phosphoric acid. It produces to give a single diastereomer of the tetrahydronaphthalene derivative under UV irradiation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-tolualdehyde was used in determination of alkenal-2,4-dinitrophenylhydrazones by HPLC by addition of phosphoric acid. It produces to give a single diastereomer of the tetrahydronaphthalene derivative under UV irradiation.

Solubility
Soluble in carbon tetrachloride, ethanol, ethyl ether, benzene. Not miscible in water.

Notes
Air Sensitive. Incompatible with air and oxidizing agents. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 529-20-4
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.151
MDL Number MFCD00003338
InChI Key BTFQKIATRPGRBS-UHFFFAOYSA-N
Synonym o-tolualdehyde, 2-tolualdehyde, 2-formyltoluene, o-toluylaldehyde, o-toluic aldehyde, o-tolylaldehyde, o-methylbenzaldehyde, benzaldehyde, 2-methyl, 2-methyl-benzaldehyde, o-methylbenazldehyde
PubChem CID 10722
ChEBI CHEBI:27434
IUPAC Name 2-methylbenzaldehyde
SMILES CC1=CC=CC=C1C=O

Specifications

Melting Point -35°C
Boiling Point 199°C to 200°C
Flash Point 67°C (152°F)
Odor Almond-like
Quantity 500 g
Beilstein 605841
Merck Index 14,9527
Formula Weight 120.15
Percent Purity 98%
Chemical Name or Material o-Tolualdehyde, Stabilized with 0.1% hydroquinone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.