missing translation for 'onlineSavingsMsg'
Learn More

o-Tolidine solution, for spectrophotometric det. of Au, Cl, according to Ellms-Hauser, Honeywell Fluka™

Product Code. 15607400 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15607400 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15607400 Supplier Honeywell-Fluka Supplier No. 89584500ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

for spectrophotometric det. of Au, Cl, according to Ellms-Hauser

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 119-93-7, 7732-18-5, 7647-01-0
Molecular Formula C14H16N2
Linear Formula C14H16N2
MDL Number MFCD00014773
UN Number UN1789
Beilstein 2210640
Density 1.015 g/cm3
Synonym o-tolidine, 3,3'-dimethylbenzidine, orthotolidine, diaminoditolyl, 2-tolidine, diaminotolyl, bianisidine, 3,3'-tolidine, 3,3'-dimethylbiphenyl-4,4'-diamine, 4,4'-bi-o-toluidine
InChI Key NUIURNJTPRWVAP-UHFFFAOYSA-N
SMILES CC1=CC(=CC=C1N)C1=CC=C(N)C(C)=C1
IUPAC Name 3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diamine
Molecular Weight (g/mol) 212.30
PubChem CID 8413
ChEBI CHEBI:34320
Formula Weight 212.29
Grade Spectrophotometry
Packaging Glass Bottle
Quantity 500 mL
Chemical Name or Material o-Tolidine solution, for spectrophotometric det. of Au, Cl, according to Ellms-Hauser
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.