missing translation for 'onlineSavingsMsg'
Learn More

o-Tolidine dihydrochloride, ACS

Product Code. 11318068
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11318068 25 g 25g
11328068 100 g 100g
11485180 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11318068 Supplier Thermo Scientific Alfa Aesar Supplier No. 036626.14
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 612-82-8 | C14H18Cl2N2 | 285.212 g/mol

o-Tolidine is employed as a colorimetric reagent for gold and free chlorine in water. Used in the synthesis of 9,9-bis(3-methyl-4-aminophenyl)fluorine. It is utilized for the detection of reducing sugars by thin layer chromatography. It is used in hair dyes, as a pharmaceutical intermediate and laboratory reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 612-82-8
Molecular Formula C14H18Cl2N2
Molecular Weight (g/mol) 285.212
MDL Number MFCD00012960
InChI Key LUKPNZHXJRJBAN-UHFFFAOYSA-N
Synonym 3,3'-dimethylbenzidine dihydrochloride, o-tolidine dihydrochloride, 3,3'-dimethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride, unii-5msk350kd8, dsstox_cid_511, dsstox_rid_75633, dsstox_gsid_20511, tolidine dihydrochloride, 1,1'-biphenyl-4,4'-diamine, 3,3'-dimethyl-, dihydrochloride, 3,3'-dimethylbenzidine.2hcl
PubChem CID 108938
IUPAC Name 4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride
SMILES CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N.Cl.Cl

Specifications

Melting Point >300°C
Color White to Cream
Odor Odorless
Quantity 25 g
UN Number UN3077
Sensitivity Air and light sensitive
Solubility Information Soluble in water,alcohol,ether and dilute acids
Formula Weight 285.22
Grade ACS
Physical Form Solid
Chemical Name or Material o-Tolidine dihydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.