missing translation for 'onlineSavingsMsg'
Learn More

o-Tolidine dihydrochloride, 97%

Product Code. p-7024038
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11413727

Brand: Thermo Scientific Alfa Aesar A18196.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 612-82-8 | C14H18Cl2N2 | 285.212 g/mol

o-Tolidine dihydrochloride is used as a sensitive colorimetric reagent for gold and for free chlorine in water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Tolidine dihydrochloride is used as a sensitive colorimetric reagent for gold and for free chlorine in water.

Solubility
Soluble in water.

Notes
Air and light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 612-82-8
Molecular Formula C14H18Cl2N2
Molecular Weight (g/mol) 285.212
MDL Number MFCD00012960
InChI Key LUKPNZHXJRJBAN-UHFFFAOYSA-N
Synonym 3,3'-dimethylbenzidine dihydrochloride, o-tolidine dihydrochloride, 3,3'-dimethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride, unii-5msk350kd8, dsstox_cid_511, dsstox_rid_75633, dsstox_gsid_20511, tolidine dihydrochloride, 1,1'-biphenyl-4,4'-diamine, 3,3'-dimethyl-, dihydrochloride, 3,3'-dimethylbenzidine.2hcl
PubChem CID 108938
IUPAC Name 4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride
SMILES CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N.Cl.Cl

Specifications

Melting Point >300°C
Odor Odorless
Quantity 25 g
UN Number UN3077
Sensitivity Air and light sensitive
Solubility Information Soluble in water.
Formula Weight 285.22
Percent Purity 97%
Chemical Name or Material o-Tolidine dihydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.