missing translation for 'onlineSavingsMsg'
Learn More

O-(tert-Butyldimethylsilyl)hydroxylamine, 90+%, Thermo Scientific Chemicals

Product Code. 11478710
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11478710 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11478710 Supplier Thermo Scientific Alfa Aesar Supplier No. B21001.03
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 41879-39-4 | Molecular Formula: C6H17NOSi | Molecular Weight: 147.293 g/mol

O-(tert-Butyldimethylsilyl)hydroxylamine (tert-butyldimethylsilyl-O-hydroxylamine)is used to induce the cleavage, during solid supported synthesis of hydroxamic acids. It also finds it uses as a chemical additive and an intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-(tert-Butyldimethylsilyl)hydroxylamine (tert-butyldimethylsilyl-O-hydroxylamine)is used to induce the cleavage, during solid supported synthesis of hydroxamic acids. It also finds it uses as a chemical additive and an intermediate.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from oxidizing agents, water/moisture and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41879-39-4
Molecular Formula C6H17NOSi
Molecular Weight (g/mol) 147.293
MDL Number MFCD00054944
InChI Key SSUCKKNRCOFUPT-UHFFFAOYSA-N
Synonym o-tert-butyldimethylsilyl hydroxylamine, o-t-butyldimethylsilyl hydroxylamine, aminoxy-tert-butyldimethylsilane, o-tert-butyldimethylsilylhydroxylamine, o-tert-butyl dimethyl silyl hydroxylamine, h2n-otbs, tbs-onh2, acmc-1asp9, hydroxylamine, tbdms derivative, o-t-butyldimethylsilylhydroxylamine
PubChem CID 4691350
IUPAC Name O-[tert-butyl(dimethyl)silyl]hydroxylamine
SMILES CC(C)(C)[Si](C)(C)ON

Specifications

Boiling Point 87°C to 90°C (40mmHg)
Melting Point 62°C to 65°C
Quantity 1 g
Sensitivity Moisture sensitive
UN Number UN3259
Solubility Information Hydrolyzes in water.
Formula Weight 147.29
Percent Purity ≥90%
Chemical Name or Material O-(tert-Butyldimethylsilyl)hydroxylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.