missing translation for 'onlineSavingsMsg'
Learn More

O-tert-Butyl-N-Fmoc-L-beta-homoserine, 95%

Product Code. 15458577
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458577 250 mg 250mg
15448577 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15458577 Supplier Thermo Scientific Alfa Aesar Supplier No. H52065.MD
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 203854-51-7 | C23H27NO5 | 397.471 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 203854-51-7
Molecular Formula C23H27NO5
Molecular Weight (g/mol) 397.471
MDL Number MFCD01862865
InChI Key SKYFRYZPXHNPOK-OAHLLOKOSA-N
Synonym fmoc-beta-hoser tbu-oh, fmoc-o-t-butyl-l-beta-homoserine, fmoc-l-beta-homoserine otbu, r-3-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy butanoic acid, 3r-4-tert-butoxy-3-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, ambotzfaa6740, fmoc-hoser tbu-oh, fmoc-??-hoser tbu-oh, fmoc-beta-homoser tbu-oh, fmoc-i 2-hoser tbu-oh
PubChem CID 2761544
IUPAC Name (3R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[(2-methylpropan-2-yl)oxy]butanoic acid
SMILES CC(C)(C)OCC(CC(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Melting Point 96°C to 98°C
Quantity 250 mg
Beilstein 8014825
Formula Weight 397.47
Percent Purity 95%
Chemical Name or Material O-tert-Butyl-N-Fmoc-L-beta-homoserine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.