missing translation for 'onlineSavingsMsg'
Learn More

O,S-Diethyl Methylphosphonothioate, TRC

Product Code. 30378193
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30378193 250 mg -
30372658 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30378193 Supplier Toronto Research Chemicals Supplier No. TRCD444460250MG
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2511-10-6
Molecular Formula C5 H13 O2 P S
Molecular Weight (g/mol) 168.19
IUPAC Name 1-[ethylsulfanyl(methyl)phosphoryl]oxyethane
SMILES CCOP(=O)(C)SCC

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C5H13O2PS/c1-4-7-8(3,6)9-5-2/h4-5H2,1-3H3
Formula Weight 168.0374
Chemical Name or Material O,S-Diethyl Methylphosphonothioate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.