missing translation for 'onlineSavingsMsg'
Learn More

O,S-Diethyl methylphosphonothioate, 97%, Thermo Scientific Chemicals

Product Code. p-7059357
Click to view available options
Quantity:
0.5 g
2 g
Unit Size:
0.5g
2g
This item is not returnable. View return policy

Product Code. 11400790

Brand: Thermo Scientific Alfa Aesar 044465.02

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

O,S-Diethyl methylphosphonothioate is used as an inhibitor of cholinesterase. It is also used in pharmaceuticals, intermediates and fine chemicals. It also serves as phosphorylating and phosphitylating agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2511-10-6
MDL Number MFCD01705977

Specifications

Density 1.094 g/mL
Boiling Point 95°C to 97°C (10 mmHg)
Molecular Formula C5H13O2PS
Refractive Index 1.474
Quantity 0.5 g
UN Number UN3278
Synonym o,s-diethyl methylphosphonothioate, o,s-diethyl methylthiophosphonate, methylphosphonothioic acid o,s-diethyl ester, osdmp, phosphonothioic acid, methyl-, o,s-diethyl ester, phosphonothioic acid, p-methyl-, o,s-diethyl ester, o-ethyl s-ethyl methylphosphonothioate, ethyl ethylsulfanyl methyl phosphinate, acmc-20ak0l, ethoxy ethylthio methyl phosphine oxide
Solubility Information Immiscible with water.
InChI Key AIMOGGQLCCAPQJ-UHFFFAOYNA-N
SMILES CCOP(=O)(C)SCC
IUPAC Name 1-[ethylsulfanyl(methyl)phosphoryl]oxyethane
Molecular Weight (g/mol) 168.19
PubChem CID 520177
Formula Weight 168.19
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material O,S-Diethyl methylphosphonothioate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.